Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009460.2 | NP_033486.1 | 5 | 402 | |
GenBank mRNA | AF033353.1 | - | 5 | 377 | |
AK002536.1 | - | 5 | 372 | ||
AK011074.1 | - | 5 | 371 | ||
AK089081.1 | - | 5 | 351 | ||
AK159366.1 | - | 5 | 373 | ||
AK220052.1 | - | 4 | 266 | ||
BC082566.1 | - | 5 | 377 | 30602751 | |
BC083158.1 | - | 5 | 375 | 6478032 |