Gene Transcripts

Gene Symbol
Luzp2
Entrez Gene ID
233271
Gene Name
leucine zipper protein 2
Gene Aliases
9330154K17Rik, D130073E01
Chromosome Location
Chr. 7: 54835200 - 55268888 on Build GRCm38
UniGene ID
Mm.387344
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178705.5 NP_848820.4 8 1291
GenBank mRNA AK032204.1 - 8 948
AK034077.1 - 8 909
AK034240.1 - 8 943
AK082126.1 - 8 1039
AY164459.1 - 8 1288
BC075646.1 - 8 921 30620475

View Full Product Details