Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033339.3 | NP_001028511.1 | 3 | 908 | |
GenBank mRNA | AK089273.1 | - | 3 | 648 | |
AK089533.1 | - | 3 | 648 | ||
AK137491.1 | - | 3 | 616 | ||
AK154458.1 | - | 3 | 910 | ||
AK171493.1 | - | 3 | 648 | ||
BC059059.1 | - | 3 | 882 | ||
BC112379.1 | - | 3 | 594 | 40059601 |