Gene Transcripts

Gene Symbol
Lrrc55
Entrez Gene ID
241528
Gene Name
leucine rich repeat containing 55
Gene Aliases
A330097E02Rik, Gm351
Chromosome Location
Chr. 2: 85188067 - 85198932 on Build GRCm38
UniGene ID
Mm.291095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033346.2 NP_001028518.1 2 819
GenBank mRNA AK134968.1 - 820
BC141221.1 - 819 9055833

View Full Product Details