Gene Transcripts

Gene Symbol
Lrit3
Entrez Gene ID
242235
Gene Name
leucine-rich repeat, immunoglobulin-like and transmembrane domains 3
Gene Aliases
Gm421
Chromosome Location
Chr. 3: 129786929 - 129808884 on Build GRCm38
UniGene ID
Mm.302549
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287224.1 NP_001274153.1 4 1973
GenBank mRNA KF954709.1 - 4 1973

View Full Product Details