Gene Transcripts

Gene Symbol
Nxf3
Entrez Gene ID
245610
Gene Name
nuclear RNA export factor 3
Gene Aliases
Gm384
Chromosome Location
Chr. X: 136072099 - 136085553 on Build GRCm38
UniGene ID
Mm.336244
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024141.4 NP_001019312.1 10, 11 1063
GenBank mRNA AB178899.1 - 10, 11 1066

View Full Product Details