Gene Transcripts

Gene Symbol
Magel2
Entrez Gene ID
27385
Gene Name
melanoma antigen, family L, 2
Gene Aliases
Mage-l2, NDNL1, nM15, ns7
Chromosome Location
Chr. 7: 62376979 - 62381640 on Build GRCm38
UniGene ID
Mm.45207
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013779.2 NP_038807.4 1 1419
GenBank mRNA AK082944.1 - 1 1392
AK086725.1 - 1 1397
BC062913.1 - 2 1304

View Full Product Details