Gene Transcripts

Gene Symbol
Rasl10b
Entrez Gene ID
276952
Gene Name
RAS-like, family 10, member B
Gene Aliases
B230331P10Rik, Gm729, VTS58635
Chromosome Location
Chr. 11: 83409142 - 83421039 on Build GRCm38
UniGene ID
Mm.66275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013386.2 NP_001013404.1 2 709
GenBank mRNA AK045997.1 - 2 384
AK158941.1 - 2 710
BC116407.2 - 2 248 40102515

View Full Product Details