Gene Transcripts

Gene Symbol
Rbp7
Entrez Gene ID
63954
Gene Name
retinol binding protein 7, cellular
Gene Aliases
1110002J23Rik, CRBP-III
Chromosome Location
Chr. 4: 149449687 - 149466462 on Build GRCm38
UniGene ID
Mm.46023
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022020.2 NP_071303.1 2 243
GenBank mRNA AF260923.1 - 2 244
AK003307.1 - 1 51
AY141971.1 - 2 253
BC028432.1 - 2 240 1348253

View Full Product Details