Gene Transcripts

Gene Symbol
Lrriq1
Entrez Gene ID
74978
Gene Name
leucine-rich repeats and IQ motif containing 1
Gene Aliases
4930503E15Rik, Gm1557
Chromosome Location
Chr. 10: 103044377 - 103236331 on Build GRCm38
UniGene ID
Mm.483817
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163559.1 NP_001157031.1 17 4015

View Full Product Details