Gene Transcripts

Gene Symbol
Tfcp2l1
Entrez Gene ID
81879
Gene Name
transcription factor CP2-like 1
Gene Aliases
1810030F05Rik, 4932442M07Rik, AA575098, Cp2l1, Crtr-1, D930018N21Rik, LBP-9, Tcfcp2l1
Chromosome Location
Chr. 1: 118627923 - 118685168 on Build GRCm38
UniGene ID
Mm.24621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023755.2 NP_076244.2 5 577
GenBank mRNA AF311309.1 - 5 572
AK143960.1 - 5 576
BC116663.1 - 5 572 40087602

View Full Product Details