Gene Transcripts

Gene Symbol
Mettl11b
Entrez Gene ID
289167
Gene Name
methyltransferase like 11B
Gene Aliases
NTM1B, RGD1564106
Chromosome Location
Chr. 13: 81987877 - 82006084 on Build Rnor_6.0
UniGene ID
Rn.225912
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_222824.7 XP_222824.5 3, 4 925
GenBank mRNA FQ213916.1 - 2, 3 670

View Full Product Details