Gene Transcripts

Gene Symbol
Vangl1
Entrez Gene ID
690366
Gene Name
VANGL planar cell polarity protein 1
Gene Aliases
Chromosome Location
Chr. 2: 204575596 - 204625835 on Build Rnor_6.0
UniGene ID
Rn.35269
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109584.1 NP_001103054.1 8 1601
XM_006233054.2 XP_006233116.1 9 1681
XM_006233055.2 XP_006233117.1 9 1644

View Full Product Details