Gene Transcripts

Gene Symbol
Pgr15l
Entrez Gene ID
296867
Gene Name
G protein-coupled receptor 15-like
Gene Aliases
RGD1562685
Chromosome Location
Chr. X: 66314416 - 66319928 on Build Rnor_6.0
UniGene ID
Rn.218317
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106583.1 NP_001100053.1 2 447

View Full Product Details