Gene Transcripts

Gene Symbol
Sass6
Entrez Gene ID
310807
Gene Name
spindle assembly 6 homolog (C. elegans)
Gene Aliases
RGD1306255
Chromosome Location
Chr. 2: 219628695 - 219660975 on Build Rnor_6.0
UniGene ID
Rn.229639
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107719.1 NP_001101189.1 3, 4 627
XM_006233227.1 XP_006233289.1 3, 4 627
XM_006233228.1 XP_006233290.1 3, 4 627

View Full Product Details