Gene Transcripts

Gene Symbol
Lrrc55
Entrez Gene ID
311171
Gene Name
leucine rich repeat containing 55
Gene Aliases
Chromosome Location
Chr. 3: 72589840 - 72604577 on Build Rnor_6.0
UniGene ID
Rn.209245
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122975.1 NP_001116447.1 1 358
XM_006234472.2 XP_006234534.1 2 1470
GenBank mRNA BC099134.1 - 1 358

View Full Product Details