Gene Transcripts

Gene Symbol
Phf20
Entrez Gene ID
311575
Gene Name
PHD finger protein 20
Gene Aliases
RGD1305020
Chromosome Location
Chr. 3: 152273269 - 152379742 on Build Rnor_6.0
UniGene ID
Rn.14290
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107795.1 NP_001101265.1 15 2448
XM_006235404.2 XP_006235466.1 16 2524
XM_008762342.1 XP_008760564.1 16 2535

View Full Product Details