Gene Transcripts

Gene Symbol
Rnft1
Entrez Gene ID
360595
Gene Name
ring finger protein, transmembrane 1
Gene Aliases
RGD1562411, Rnft1-ps1
Chromosome Location
Chr. 10: 73807454 - 73821178 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750899.3 XP_003750947.1 6 1022
XM_006247119.2 XP_006247181.1 6 1248

View Full Product Details