Gene Transcripts

Gene Symbol
Xylt1
Entrez Gene ID
64133
Gene Name
xylosyltransferase 1
Gene Aliases
XYLT-1, Xt1
Chromosome Location
Chr. 1: 186939698 - 187264758 on Build Rnor_6.0
UniGene ID
Rn.162460
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006230151.2 XP_006230213.2 5 1068
GenBank mRNA AJ295748.1 - 2 574

View Full Product Details