Gene Transcripts

Gene Symbol
Prl2a1
Entrez Gene ID
116474
Gene Name
Prolactin family 2, subfamily a, member 1
Gene Aliases
Prlpm
Chromosome Location
Chr. 17: 38900411 - 38906503 on Build Rnor_6.0
UniGene ID
Rn.17940
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053791.2 NP_446243.1 4 445
GenBank mRNA AF226608.1 - 4 443
BC097296.1 - 4 445 7445776
DQ329279.1 - 4 443

View Full Product Details