Gene Transcripts

Gene Symbol
Lsm2
Entrez Gene ID
684148
Gene Name
LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
G7b
Chromosome Location
Chr. 20: 4966817 - 4970567 on Build Rnor_6.0
UniGene ID
Rn.216424
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004073.1 NP_001004073.1 4 241
NM_001165922.1 NP_001159394.1 4 338

Gene Symbol
LOC100361344
Entrez Gene ID
100361344
Gene Name
hypothetical LOC100361344
Gene Aliases
Chromosome Location
Chr. 19: 44396323 - 44408628 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772593.1 XP_008770815.1 2 238

View Full Product Details