Gene Transcripts

Gene Symbol
Olr62
Entrez Gene ID
293201
Gene Name
olfactory receptor 62
Gene Aliases
ratchr1-160722887-160723849_ORF
Chromosome Location
Chr. 1: 167874539 - 167875501 on Build Rnor_6.0
UniGene ID
Rn.141917
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000129.1 NP_001000129.1 1 377

View Full Product Details