Gene Transcripts

Gene Symbol
Ticam1
Entrez Gene ID
363328
Gene Name
toll-like receptor adaptor molecule 1
Gene Aliases
Chromosome Location
Chr. 9: 10780236 - 10790280 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766802.1 XP_008765024.1 2 582

View Full Product Details