Gene Transcripts

Gene Symbol
Lyzl1
Entrez Gene ID
364745
Gene Name
lysozyme-like 1
Gene Aliases
RGD1559869
Chromosome Location
Chr. 17: 56255613 - 56269245 on Build Rnor_6.0
UniGene ID
Rn.124844
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108882.1 NP_001102352.1 3 298

View Full Product Details