Gene Transcripts

Gene Symbol
Chd4
Entrez Gene ID
117535
Gene Name
chromodomain helicase DNA binding protein 4
Gene Aliases
Mi-2
Chromosome Location
Chr. 4: 157612540 - 157645660 on Build Rnor_6.0
UniGene ID
Rn.63247
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006237395.2 XP_006237457.1 29 4780
XM_006237396.2 XP_006237458.1 29 4780
XM_006237397.2 XP_006237459.1 30 4695
XM_006237398.2 XP_006237460.1 30 4692
XM_006237399.2 XP_006237461.1 30 4674
XM_006237400.2 XP_006237462.1 30 4656
XM_006237401.2 XP_006237463.1 30 4695
XM_232354.8 XP_232354.5 30 4674

View Full Product Details