Gene Transcripts

Gene Symbol
Nkx2-4
Entrez Gene ID
366213
Gene Name
NK2 homeobox 4
Gene Aliases
Chromosome Location
Chr. 3: 141303357 - 141305489 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_345445.7 XP_345446.3 2 771

View Full Product Details