Gene Transcripts

Gene Symbol
Psme4
Entrez Gene ID
498433
Gene Name
proteasome (prosome, macropain) activator subunit 4
Gene Aliases
Chromosome Location
Chr. 14: 115165915 - 115269041 on Build Rnor_6.0
UniGene ID
Rn.163607
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025140.2 NP_001020311.2 43 5294
XM_008770467.1 XP_008768689.1 44 5610
GenBank mRNA CK600326.1 - 5 564

View Full Product Details