Gene Transcripts

Gene Symbol
Atg9b
Entrez Gene ID
499973
Gene Name
autophagy related 9B
Gene Aliases
RGD1560887
Chromosome Location
Chr. 4: 7314369 - 7321239 on Build Rnor_6.0
UniGene ID
Rn.228194
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191968.1 NP_001178897.1 6 1262
XM_006235960.2 XP_006236022.1 6 1262

View Full Product Details