Gene Transcripts
- Gene Symbol
- RT1-CE6
- Entrez Gene ID
- 415073
- Gene Name
- RT1 class I, locus CE6
- Gene Aliases
- Chromosome Location
- Chr. 20: 4676851 - 4677801 on Build Rnor_6.0
- UniGene ID
- Rn.233151
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002155.1 |
|
1 |
466 |
|
View Full Product Details