Gene Transcripts
- Gene Symbol
- Rmrp
- Entrez Gene ID
- 19782
- Gene Name
- RNA component of mitochondrial RNAase P
- Gene Aliases
- 1110032O22Rik, Rmrpr
- Chromosome Location
- Chr. 4: 43492785 - 43493059 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001460.1 |
|
LIT3323 RMRP |
1 |
44 |
|
View Full Product Details