Gene Transcripts
- Gene Symbol
- Mx1
- Entrez Gene ID
- 17857
- Gene Name
- MX dynamin-like GTPase 1
- Gene Aliases
- AI893580, Mx, Mx-1
- Chromosome Location
- Chr. 16: 97447035 - 97462906 on Build GRCm38
- UniGene ID
- Mm.33996
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ134499.1 |
- |
1 |
75 |
|
Chr. 16 - 97456292 - 97462901 [-] |
GenBankmRNA |
CJ134894.1 |
- |
1 |
75 |
|
Chr. 16 - 97456292 - 97462901 [-] |
GenBankmRNA |
BI853349.1 |
- |
1 |
63 |
|
Chr. 16 - 97455164 - 97462889 [-] |
View Full Product Details