Gene Transcripts

Gene Symbol
RAI2
Entrez Gene ID
10742
Gene Name
retinoic acid induced 2
Gene Aliases
Chromosome Location
Chr. X: 17800049 - 17861346 on Build GRCh38
UniGene ID
Hs.446680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033348.1 1 240
NR_033349.1 1 240
XM_011545439.2 XP_011543741.1 1 202
XM_011545440.2 XP_011543742.1 1 202
XM_011545441.2 XP_011543743.1 1 202
GenBank mRNA BC027937.1 - 1 120
DA377448.1 - 1 185
DR005700.1 - 1 246

View Full Product Details