Gene Transcripts

Gene Symbol
MTMR9LP
Entrez Gene ID
339483
Gene Name
myotubularin related protein 9-like, pseudogene
Gene Aliases
MTMR9L
Chromosome Location
Chr. 1: 32231660 - 32241710 on Build GRCh38
UniGene ID
Hs.471067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026850.1 6 1940
GenBank mRNA AK055560.1 - 7 1672
AK096109.1 - 5 1492
AL833686.1 - 8 2240
BC016171.1 - 3 785
BC041632.1 - 3 595


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB549336.1 - 1 73 Chr. 1 - 32231661 - 32232184 [-]
GenBankmRNA BG761082.1 - 3 596 Chr. 1 - 32231912 - 32240232 [-]
GenBankmRNA BM747778.1 - 3 210 Chr. 1 - 32231697 - 32234981 [-]
GenBankmRNA BM833152.1 - 3 210 Chr. 1 - 32231698 - 32234981 [-]
GenBankmRNA AA251929.1 - 3 278 Chr. 1 - 32232052 - 32235028 [-]
GenBankmRNA AW967629.1 - 3 278 Chr. 1 - 32231747 - 32235028 [-]
GenBankmRNA CN430607.1 - 3 582 Chr. 1 - 32231943 - 32240219 [-]

View Full Product Details