Gene Transcripts

Gene Symbol
MYH16
Entrez Gene ID
84176
Gene Name
myosin heavy chain 16 pseudogene
Gene Aliases
MHC20, MYH16P, MYH5
Chromosome Location
Chr. 7: 99273301 - 99297972 on Build GRCh38
UniGene ID
Hs.621401
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002147.2 14, 15 2060
GenBank mRNA AK025690.1 - 14, 15 2060

View Full Product Details