Gene Transcripts
- Gene Symbol
- MYH16
- Entrez Gene ID
- 84176
- Gene Name
- myosin heavy chain 16 pseudogene
- Gene Aliases
- MHC20, MYH16P, MYH5
- Chromosome Location
- Chr. 7: 99273301 - 99297972 on Build GRCh38
- UniGene ID
- Hs.621401
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002147.2 |
|
10, 11 |
1383 |
|
GenBank mRNA |
AK025690.1 |
- |
10, 11 |
1383 |
|
View Full Product Details