Gene Transcripts
- Gene Symbol
- SCARNA12
- Entrez Gene ID
- 677777
- Gene Name
- small Cajal body-specific RNA 12
- Gene Aliases
- U89
- Chromosome Location
- Chr. 12: 6967337 - 6967606 on Build GRCh38
- UniGene ID
- Hs.689636
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003010.1 |
|
1 |
98 |
|
GenBank mRNA |
AY077739.1 |
- |
1 |
98 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG818346.1 |
- |
1 |
112 |
|
Chr. 12 - 6966777 - 6967620 [-] |
View Full Product Details