Gene Transcripts
- Gene Symbol
- BRD7P3
- Entrez Gene ID
- 23629
- Gene Name
- bromodomain containing 7 pseudogene 3
- Gene Aliases
- BP75
- Chromosome Location
- Chr. 6: 118501373 - 118503846 on Build GRCh38
- UniGene ID
- Hs.648050
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002730.2 |
|
1 |
1971 |
|
GenBank mRNA |
BC033937.1 |
- |
1 |
725 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG714720.1 |
- |
1 |
725 |
|
Chr. 6 - 118502621 - 118503354 [+] |
View Full Product Details