Gene Transcripts
- Gene Symbol
- PI4KAP1
- Entrez Gene ID
- 728233
- Gene Name
- phosphatidylinositol 4-kinase alpha pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 22: 18533864 - 18548828 on Build GRCh38
- UniGene ID
- Hs.592456
- Species
- Homo sapiens
- Gene Symbol
- PI4KA
- Entrez Gene ID
- 5297
- Gene Name
- phosphatidylinositol 4-kinase alpha
- Gene Aliases
- PI4K-ALPHA, PIK4CA, PMGYCHA, pi4K230
- Chromosome Location
- Chr. 22: 20707691 - 20858812 on Build GRCh38
- UniGene ID
- Hs.529438
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095811.1 |
- |
11 |
1208 |
|
AK124692.1 |
- |
8 |
2133 |
|
- Gene Symbol
- PI4KAP2
- Entrez Gene ID
- 375133
- Gene Name
- phosphatidylinositol 4-kinase alpha pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 22: 21472998 - 21517491 on Build GRCh38
- UniGene ID
- Hs.448225
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK297163.1 |
- |
6 |
720 |
|
AK299734.1 |
- |
14 |
1616 |
|
BQ638971.1 |
- |
1 |
466 |
|
View Full Product Details