Gene Transcripts

Gene Symbol
MSTO2P
Entrez Gene ID
100129405
Gene Name
misato family member 2, pseudogene
Gene Aliases
MSTO2
Chromosome Location
Chr. 1: 155745768 - 155750688 on Build GRCh38
UniGene ID
Hs.656361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024117.2 9, 10 1071
GenBank mRNA BC099909.1 - 9, 10 1021

Gene Symbol
MSTO1
Entrez Gene ID
55154
Gene Name
misato 1, mitochondrial distribution and morphology regulator
Gene Aliases
LST005, MST
Chromosome Location
Chr. 1: 155610170 - 155614967 on Build GRCh38
UniGene ID
Hs.656547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011509709.2 XP_011508011.1 10, 11 1136
GenBank mRNA BC070067.1 - 7, 8 1625

View Full Product Details