Gene Transcripts
- Gene Symbol
- RPS15AP10
- Entrez Gene ID
- 728963
- Gene Name
- ribosomal protein S15a pseudogene 10
- Gene Aliases
- RPS15A_4_42
- Chromosome Location
- Chr. 1: 45645772 - 45646685 on Build GRCh38
- UniGene ID
- Hs.675157
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026768.1 |
|
1 |
347 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX481330.1 |
- |
1 |
351 |
|
Chr. 1 - 45646020 - 45646688 [-] |
GenBankmRNA |
CB162364.1 |
- |
1 |
115 |
|
Chr. 1 - 45645857 - 45646443 [-] |
GenBankmRNA |
AJ705415.1 |
- |
1 |
217 |
|
Chr. 1 - 45646056 - 45646534 [-] |
GenBankmRNA |
BG654885.1 |
- |
1 |
40 |
|
Chr. 1 - 45645815 - 45646374 [-] |
GenBankmRNA |
CA843117.1 |
- |
1 |
39 |
|
Chr. 1 - 45645772 - 45646374 [-] |
View Full Product Details