Gene Transcripts
- Gene Symbol
- NDUFB11
- Entrez Gene ID
- 54539
- Gene Name
- NADH:ubiquinone oxidoreductase subunit B11
- Gene Aliases
- CI-ESSS, ESSS, LSDMCA3, NP17.3, Np15, P17.3
- Chromosome Location
- Chr. X: 47142216 - 47145210 on Build GRCh38
- UniGene ID
- Hs.521969
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN301578.1 |
- |
1 |
156 |
|
Chr. X - 47144610 - 47145029 [-] |
GenBankmRNA |
BM839821.1 |
- |
1 |
188 |
|
Chr. X - 47144610 - 47145089 [-] |
GenBankmRNA |
BE781599.1 |
- |
1 |
96 |
|
Chr. X - 47142316 - 47144997 [-] |
GenBankmRNA |
CN301576.1 |
- |
1 |
172 |
|
Chr. X - 47142621 - 47145073 [-] |
View Full Product Details