Gene Transcripts
- Gene Symbol
- Mirg
- Entrez Gene ID
- 100040724
- Gene Name
- miRNA containing gene
- Gene Aliases
- 2810474H01Rik, AI852992, Gm2928, Meg9, mir-410
- Chromosome Location
- Chr. 12: 109734981 - 109749458 on Build GRCm38
- UniGene ID
- Mm.260543
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ517767.1 |
- |
LIT1749 Mirg |
1 |
14 |
|
View Full Product Details