Gene Transcripts
A Gene Symbol is not available because the non-coding siRNA was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID. Non-coding siRNAs are designed to RefSeq (NR_) or to curated sequences from RNAdb represented by GenBank RNA or ESTdb accessions.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AI970466.1 |
- |
1 |
185 |
|
Chr. 11 - 2675322 - 2675845 [+] |
GenBankmRNA |
AJ006345.3 |
- |
2 |
230538 |
|
Chr. 11 - 2444100 - 2849207 [+] |
GenBankmRNA |
AI989662.1 |
- |
1 |
185 |
|
Chr. 11 - 2675322 - 2675751 [+] |
GenBankmRNA |
H88273.3 |
- |
1 |
178 |
|
Chr. 11 - 2675329 - 2675656 [+] |
GenBankmRNA |
AA029452.1 |
- |
1 |
187 |
|
Chr. 11 - 2675321 - 2675718 [+] |
GenBankmRNA |
DB351126.1 |
- |
1 |
185 |
|
Chr. 11 - 2675322 - 2675842 [+] |
View Full Product Details