Gene Transcripts
- Gene Symbol
- NAPA
- Entrez Gene ID
- 8775
- Gene Name
- NSF attachment protein alpha
- Gene Aliases
- SNAPA
- Chromosome Location
- Chr. 19: 47487634 - 47515258 on Build GRCh38
- UniGene ID
- Hs.126938
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY927608.1 |
- |
LIT3598 AY927608 |
1, 2 |
196 |
|
View Full Product Details