Gene Transcripts
- Gene Symbol
- MAP3K13
- Entrez Gene ID
- 9175
- Gene Name
- mitogen-activated protein kinase kinase kinase 13
- Gene Aliases
- LZK, MEKK13, MLK
- Chromosome Location
- Chr. 3: 185282938 - 185489094 on Build GRCh38
- UniGene ID
- Hs.591306
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC026249.1 |
- |
1 |
1512 |
|
View Full Product Details