Gene Transcripts
- Gene Symbol
- OCR1
- Entrez Gene ID
- 100128298
- Gene Name
- ovarian cancer-related protein 1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.679412
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF314543.1 |
- |
1 |
42 |
|
View Full Product Details