Gene Transcripts
- Gene Symbol
- MESP1
- Entrez Gene ID
- 55897
- Gene Name
- mesoderm posterior bHLH transcription factor 1
- Gene Aliases
- bHLHc5
- Chromosome Location
- Chr. 15: 89734772 - 89751309 on Build GRCh38
- UniGene ID
- Hs.447531
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL357535.1 |
- |
1 |
1330 |
|
View Full Product Details