Gene Transcripts
- Gene Symbol
- HEMK1
- Entrez Gene ID
- 51409
- Gene Name
- HemK methyltransferase family member 1
- Gene Aliases
- HEMK, MTQ1
- Chromosome Location
- Chr. 3: 50569152 - 50589050 on Build GRCh38
- UniGene ID
- Hs.517987
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF074992.1 |
- |
1 |
381 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX113747.1 |
- |
1 |
378 |
|
Chr. 3 - 50588537 - 50589050 [+] |
View Full Product Details