Gene Transcripts

Gene Symbol
RRP1
Entrez Gene ID
8568
Gene Name
ribosomal RNA processing 1
Gene Aliases
D21S2056E, NNP-1, NOP52, RRP1A
Chromosome Location
Chr. 21: 43789537 - 43804102 on Build GRCh38
UniGene ID
Hs.110757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK026354.1 - 1 101


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AW901603.1 - 1 286 Chr. 21 - 43803045 - 43803536 [+]
GenBankmRNA AA477351.1 - 1 288 Chr. 21 - 43803033 - 43803399 [+]
GenBankmRNA CA388865.1 - 1 259 Chr. 21 - 43803058 - 43803399 [+]
GenBankmRNA BQ723160.1 - 181 Chr. - - [-]
GenBankmRNA BP263582.1 - 1 101 Chr. 21 - 43803216 - 43803797 [+]
GenBankmRNA CR745199.1 - 1 284 Chr. 21 - 43803033 - 43803454 [+]
GenBankmRNA BF363881.1 - 1 239 Chr. 21 - 43803083 - 43803555 [+]

View Full Product Details