Gene Transcripts
- Gene Symbol
- OAS1
- Entrez Gene ID
- 4938
- Gene Name
- 2'-5'-oligoadenylate synthetase 1
- Gene Aliases
- E18/E16, IFI-4, OIAS, OIASI
- Chromosome Location
- Chr. 12: 112906777 - 112932190 on Build GRCh38
- UniGene ID
- Hs.524760
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123528.1 |
- |
1 |
1588 |
|
View Full Product Details